Utility Shear with Footplate, Walter Stern, Inc.
Supplier: Walter Stern
310-341
57252-150EA 32.19 USD
57252-150
Utility Shear with Footplate, Walter Stern, Inc.
Scissors
This multipurpose shear is designed with stainless steel blades and plastic handle.
  • Blade and handle are autoclavable upto 120 °C
Bottom blade is equipped with footplate to aid in cutting between bandages and skin as well as other areas where safety must be considered.