General-Purpose Burn Kit, Honeywell Safety
Supplier: Honeywell Safety Products
019727-0014L
57100-340EA 440.13 USD
57100-340
General-Purpose Burn Kit, Honeywell Safety
First Aid Supplies Burn Care Supplies
Contains both dry and wet dressings.
Dry dressings protect against contamination and wet dressings help draw heat out of the burn.