Holder for Bench Top Reciprocating Shaker, Erlenmeyer Flask, Glas-Col
Supplier: Glas-Col
099AEF521
57020-289EA 1029.69 USD
57020-289
Holder for Bench Top Reciprocating Shaker, Erlenmeyer Flask, Glas-Col
Supports Flask Supports
This flask holder lets you shake one to four Erlenmeyer flasks at one time on our Bench Top Shaker.
  • Easy adjustment accomodates one to four flasks
  • Locking knob secures holder in optimum positions for loading and effective shaking
  • Holder can be rotated 180 degrees while mounted on shaker
  • Two flask holder can be shaken at once
Uncapped flasks can be shaken back and forth gently or you can close the flasks when a more vigorous back and forth motion is needed.
Ordering information: Accomodates three 500 mL flasks or four 125 mL to 250 L flasks.
Delivery information: 7-10 days after order.