Chemware® Dipper, Teflon® PFA, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
D1069102
56925-300EA 113.44 USD
56925-300
Chemware® Dipper, Teflon® PFA, Saint-Gobain Performance Plastics
Samplers Sample Dippers
Use for ultra-pure applications and with aggressive media
Handle hooks over edge of container, eliminating dripping of corrosive fluids onto surfaces. The 6.4 cm (21/2") diameter allows for use with containers having at least a 7.6 cm (3") opening and a capacity ranging from 300 to 1000ml.