SP Bel-Art Sterileware® Sampling Knife, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H36965-0000
56924-478CS 498.38 USD
56924-478
SP Bel-Art Sterileware® Sampling Knife, Bel-Art Products, a part of SP
Knives
Ideal for cutting small samples from fibrous pieces, this sterile polystyrene plastic sampler knife has a blade that is serrated on one edge and smooth on the other.
  • Polystyrene plastic construction
  • Sterile
  • Blade is serrated on one edge and smooth on the other
  • Sterilized via gamma irradiation
  • Lot stamped for accurate lot tracing
The serrated edge is capable of cutting seafood, meat, poultry, vegetables, and other fibrous materials. Individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization.