Sample Storage Sets, Cork Closure, Cargille Laboratories
Supplier: Cargille Laboratories
36130
56780-006EA 313.59 USD
56780-006
Sample Storage Sets, Cork Closure, Cargille Laboratories
Vials
Most economical, straight-sided, flat-bottom, glass shell vials with high quality cork closures.
  • Alpha-numerically indexed compartments for accurate and rapid retrieval and refilling of samples
  • Requires little space for storing hundreds or thousands of items
  • Boxes are constructed with a moisture and abrasive-resistant exterior covering
  • Suitable for incubator, refrigerator, deep-freeze and controlled atmosphere applications
Ideally suited for storing small parts, geological specimens, soil and other types of samples not requiring chemical resistant closures.
Ordering information: Each set includes a partitioned box, vials, closures, gummed labels, reference chart, an exterior card holder, and card.