Nalgene® Sample Bags, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
6255-0508 6255-0613 6255-0913 6255-0918
56766-049CS 503.65 USD
56766-049 56766-071 56766-107 56766-130
Nalgene® Sample Bags, Low-Density Polyethylene, Thermo Scientific
Bags Sample Bags
Transparent LDPE sample bags with strong, 4 mil walls are ideal for collecting, storing, sealing and protecting specimens, labware and small parts.
Sturdy 'zipper' locks securely and keeps dust out. Not recommended for storing liquids.