Sample Splitter, Humboldt
Supplier: Avantor
56723-250EA 1665.87 USD
56723-250
Sample Splitter, Humboldt
Samplers Solid Samplers
For rapid, even splitting of cements, ores and other dry samples.
Material poured into top is divided by fourteen 1.9cm (3/4") chutes that discharge into receiving pans. Constructed of stainless steel throughout. Dimensions: 17.1x 37.5cm (63/4x143/4"). Shipping weight: 9.5kg (21lbs).
Ordering information: Supplied complete with four pans and cleaning brush.