Jones Type Sample Splitter
Supplier: Avantor
56720-060EA 1819.52 USD
56720-060 56720-081
Jones Type Sample Splitter
Samplers Solid Samplers
Used for ASTM C136, D421, D271, D492 methods for rapid, even splitting of samples of aggregates, cement, road material, and other dry materials.
Sample material poured into the main hopper unit is divided by six riffles into two equal parts by a series of chutes that discharge in opposite directions into two receiving pans. By repeating the operation with reduced sample, the sample can be “cut down” to quarters, eighths, etc.
Ordering information: Choose from two sizes. Both include stainless steel hopper and chutes, four sample pans, scoop and cleaning brush.