8271 P95 Particulate Respirators, 3M™
Supplier: 3M
7000002047
56621-934CS 845.86 USD
56621-934
8271 P95 Particulate Respirators, 3M™
Respirators Disposable Respirators Particulate Respirators
Respirator provides protection against certain solid and liquid particles, including those containing oil.
  • Provides protection against certain solid and liquid particles, including those containing oil
  • Conformable "M" nose clip for an improved fit
  • Cool Flow® exhalation valve to reduce heat and moisture inside the respirator
  • Face seal provides comfort and fit over broad range of face sizes
  • If used in environments containing oil aerosols, limit use to 40 hours or 30 days
  • Provides minimum filtration efficiency of 95%
Suggested uses include grinding,More Product Information