8247 R95 Respirator, 3M™
Supplier: 3M
8247
56621-932CS 1247.6 USD
56621-932
8247 R95 Respirator, 3M™
Respirators Disposable Respirators Particulate Respirators
This respirator provides worker protection against certain oil and non-oil based aerosols including those with nuisance levels of organic vapors such as solvents, degreasers, and resins.
  • Protects against certain oil and non-oil based aerosols
  • If oil aerosols are present, respirator may be limited to 8 hours of continuous or intermittent use
  • Provides minimum filtration efficiency of 95%
(Nuisance level acid gas refers to concentrations not exceeding the OSHA PEL or applicable government occupational exposure limits, whichever is lower.) If oil aerosols are present, respirator may be limited to 8 hours of continuous or intermittent use. Provides minimum filtration efficiency of 95%.
Caution: Must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.