PIG® Rip-and-Fit® Absorbent Mat Roll in Dispenser Box, New Pig
Supplier: New Pig
MAT242
56620-712EA 111.23 USD
56620-712
PIG® Rip-and-Fit® Absorbent Mat Roll in Dispenser Box, New Pig
Spill Control Materials
Extra perforations allow you to easily rip this PIG Mat and fit it to your application without cutting.
  • Multiple perforations allow fitting to any space
  • Perforated every 33" across width and every 10" across length
  • Thermally bonded to add strength and accelerate wicking
  • Each dispenser absorbs 30.3L (8gal.) of oils, coolants, solvents, and water
No matter how you rip it, this mat features the proven eight-layer, 100% polypropylene construction of PIG Mat
Absorbs and retains oils, coolants, solvents and water. Dark gray color hides grime so mat stays on the job longer. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Dispenser box makes it easy to take, carry and store your mat while protecting it from dirt and moisture. Perforated every 3.75" across width and every 10" across length.
Heavyweight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups.
Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave behind liquids or fiber residue.