PIG® Un-Perforated Absorbent Mat Rolls, New Pig
Supplier: New Pig
MAT211
56620-580PK 173.13 USD
56620-580
PIG® Un-Perforated Absorbent Mat Rolls, New Pig
Spill Control Materials
Eight layers of 100% polypropylene are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated
Absorbs and retains oils, coolants, solvents and water. Dark gray color hides grime so mat stays on the job longer. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Can be wrung out and incinerated after use to reduce waste or for fuels blending. Un-perfed rolls can be cut to fit your exact workspace for soaking up leaks and overspray.
Heavyweight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups.
Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind