PIG® Spill Kit in 65-Gallon High-Visibility Container, New Pig
Supplier: New Pig
KIT263
56620-528EA 628.62 USD
56620-528
PIG® Spill Kit in 65-Gallon High-Visibility Container, New Pig
Spill Control Materials
Lift-out, prepacked baskets speed access and guard contents from UV
Lime green drum is easy to locate. Lightweight, 100% polyethylene drum resists chemicals and keeps contents clean and dry. Large, color-coded label makes identification fast and accurate. Nonthreaded lid lifts off for easy access (not rated for shipping). Ledges molded into container make kit easy to move by forklift. Tamperproof seal helps prevent pilfering of spill response supplies. Temporary disposal bags with ties help make cleanup faster.
No Wheels. Not UN rated.
PIG Blue Socks stop spreading spills; PIG Mat and Pillows absorb; wipers clean surfaces and hands