PIG® You-Supply-the-Drum Spill Kit, New Pig
Supplier: New Pig
KIT205
56620-504EA 350.34 USD
56620-504
PIG® You-Supply-the-Drum Spill Kit, New Pig
Spill Control Materials
Contents packed in two easy-to-lift bags for quick response with socks on top of pads and pillows
Temporary disposal bags with ties help make cleanup faster. Includes "Emergency Spill Kit" decals for easy identification.
PIG Blue Socks stop spreading spills; PIG Mat and Pillows absorb quickly.