PIG® HazMat Economy Spill Kits in Duffel Bag, New Pig
Supplier: New Pig
KIT323
56620-492EA 753.66 USD
56620-492
PIG® HazMat Economy Spill Kits in Duffel Bag, New Pig
Spill Control Materials
Just open kit, use the absorbents, then throw them in the temporary disposal bags included
  • Rugged duffel bag is packed with four kits
  • Absorb spills, then keep the bag
  • Makes a great vehicle spill kit
PIG HazMat Socks stop spreading spills; PIG HazMat Pads absorb quickly. PIG HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide.