PIG® Gray Ham-O® Poly-Back Absorbent Mat Roll, New Pig
Supplier: New Pig
MAT132
56620-420EA 607.01 USD
56620-420
PIG® Gray Ham-O® Poly-Back Absorbent Mat Roll, New Pig
Spill Control Materials
Exclusive "hamouflage" top layer saves on product and disposal costs; absorbed grime is invisible, so you'll leave mat in place longer and save money
Tough, spunbond top layer stands up to heavy foot traffic; won't rip, tear, fray or create a trip hazard, even when saturated. Under the Ham-O top layer is the proven eight-layer, 100% polypropylene construction of PIG Mat. Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Absorbs and retains oils, coolants, solvents and water. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Can be wrung out and incinerated after use to reduce waste or for fuels blending. 5-mil poly backing prevents absorbed liquids from reaching floors or counters and provides added traction for safe footing. Place over a spill with poly-back side up to suppress potentially harmful, flammable or irritating vapors. Rolls are ideal as a work surface for repairs or maintenance - poly backing prevents grease and oil from seeping through.
Heavyweight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups.
Fast-wicking "diaper action" transfers liquids from the top layer into the absorbent inner layers; spills disappear in seconds