PIG® MRO Economy Spill Kit, New Pig
Supplier: New Pig
KIT242
56620-300EA 95.47 USD
56620-300
PIG® MRO Economy Spill Kit, New Pig
Spill Control Materials
Maintenance, repair, and operation (MRO) spill kit is packed inside a heat-sealed disposal bag and is small enough to be placed anywhere in a facility or on a transport truck
  • Helps facilities comply with 40 CFR 263.30(a) safety standard
Safety-yellow bag is highly visible, labeled, and may also be reused for temporary disposal of saturated absorbents. Kit contents absorb up to 18.9L (5gal.) of nonaggressive liquids such as oils, coolants, solvents, and water.
Ordering information: Kit contains three 7.6dia.x121.9Lcm (3x48") blue socks, twelve 50.8Lx40.6Wcm (20x16") universal mat pads, and two temporary disposal bags and ties.