Uninsulated Bonding/Grounding Wires, New Pig
Supplier: New Pig
DRM495
56620-206EA 129.35 USD
56620-206 56620-208
Uninsulated Bonding/Grounding Wires, New Pig
Grounders Grounding Wires
Reduces static sparks generated by the movement and flow of flammable liquids
  • Reduces static sparks generated by the movement and flow of flammable liquids
  • Helps you meet 29 CFR 1910.106 (e)(6)(ii) for grounding and bonding when transferring Class I flammables
Helps you meet 29 CFR 1910.106 (e)(6)(ii) for grounding and bonding when transferring Class I flammables.