PIG® HazMat Chemical Absorbent Pulp, New Pig
Supplier: New Pig
HA8010
56620-186CS 153.55 USD
56620-186
PIG® HazMat Chemical Absorbent Pulp, New Pig
Spill Control Materials
Packing the power of PIG HazMat in a loose absorbent, this pulp can be used in a variety of applications
  • Absorbs and retains most acids, bases and unknown liquids
  • Pink color is easily distinguishable to assure workers use the correct pulp during a spill emergency
  • Can be incinerated after use to reduce waste
Absorbs the widest range of acids, bases and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide. Chemical-resistant pulp won't degrade or cause a dangerous reaction upon contact with corrosive spills. Excellent for difficult to reach spaces or final cleanup