PIG® Outdoor Pan, New Pig
Supplier: New Pig
PAN401
56620-092CS 154.88 USD
56620-092
PIG® Outdoor Pan, New Pig
Drum Storage
Absorbs and retains oils and oil-based liquids, including lubricants and fuels, without taking in a drop of water.
  • Mesh top lets liquid pass into absorbent filler housed in a chemical-resistant pan
  • Rainwater passes through slits in side of pan to avoid puddling
  • Low-profile pan fits in tight spaces and won't tip over
Includes PIG Oil-Only Mat to absorb any additional oily discharge and protect the ground.