Miller Cross-Arm Anchorage Strap, Honeywell Safety
Supplier: Honeywell Safety Products
8183/6FTGN
56619-164EA 124.76 USD
56619-164
Miller Cross-Arm Anchorage Strap, Honeywell Safety
Fall Protection Fall Lanyards and Ropes
For securing to I-beams and other structures
  • 5.1cm (2") D-ring that slips through 7.6cm (3") D-ring to form secure attachment point for lanyards and other connecting devices
Strap features a 5.1cm (2") D-ring that slips through a 7.6cm (3") D-ring to form a secure attachment point for lanyards and other connecting devices. (Sperian 8183/6FTGN)
Maximum Capacity: 181 kg (400 lbs.)