Biohazard Waste Can Liner Bags, Justrite®
Supplier: Justrite
05901
56617-834PK 138.24 USD
56617-834
Biohazard Waste Can Liner Bags, Justrite®
Bags Waste Disposal Bags
For safe, temporary collection of biological waste or contaminated materials. Red color is easily identifiable as biohazard bags.
  • 15 Gallon (56,8 l) capacity bag fits Justrite Biohazard cans of 6 and 10 gallon sizes
  • High density PE of 11 microns thickness provides secure handling to avoid punctures and tears
  • Tubular construction with a single side seal and star seal bottom provide a stable, leak proof bag
  • Specifically labeled for Biohazard material handling
Printing includes English, Spanish and French translation along with the international recognizable biohazard symbol.