Lab Pack, UN and DOT, Eagle Manufacturing
Supplier: Eagle Manufacturing
1652
56617-331EA 74.69 USD
56617-331
Lab Pack, UN and DOT, Eagle Manufacturing
Mailers and Shippers
Suitable for use with hazardous materials, Packing Groups I, II, and III.
Constructed of blow-molded high-density polyethylene with UV inhibitors. Chemical-resistant to most materials. Incinerates completely. Nestable for easy storage. Handles are suitable for forklifting.
UN certified to HM-181. Meets DOT 49 CFR 173.12(b). UN Marking: 1H2/x40/S.
Holds 18.9L (5gal.) pails or smaller.