ESD Safe Sports Bottle, R & R Lotion
Supplier: R & R Lotion
SB-32-ESD
56616-120CS 492.8 USD
56616-120
ESD Safe Sports Bottle, R & R Lotion
Bottles Spray Bottles
Antistatic dissipative, ESD safe sports bottle.I.C.
  • Water/sports bottle in an ESD safe container
  • Designed for use around static sensitive electronics
  • Will not hold an electric charge
The ESD Safe Sports Bottle allows workers to keep drinks handy in static sensitive environments. Bottles feature a pull-top cap for workers on the go. These containers offer an average surface resistivity of 10⁹ to 10¹⁰. All blue solvent dispensers will dissipate a charge of +/- 5000 volts in less than 2 s at 40 percent relative humidity.