ESD Safe Water Bottle, R & R Lotion
Supplier: R & R Lotion
WB-8-ESD
56614-154EA 24.81 USD
56614-154
ESD Safe Water Bottle, R & R Lotion
Bottles Spray Bottles
The ESD safe water bottle is equipped with a cone spout for efficient dispensing.
Cleanroom blue static dissipative containers have an average surface resistivity of 10⁹ to 10¹⁰. All blue solvent dispensers will dissipate a charge of +/- 5000 volts in less than 2 s at 40 percent relative humidity. By their chemical nature, they will not have any chemical reactivity with solvents such as TCE or alcohol. Bottles help to meet ANSI/ESDA S20.20.