UltraFit™ Nylon Glove Liners, Erie Scientific (Thermo Scientific)
Supplier: Epredia
51002-12-001 51004-12-001 51001-12-001 51003-12-001
56613-472CS 767.16 USD
56613-472 56613-474 56614-284 56614-288
UltraFit™ Nylon Glove Liners, Erie Scientific (Thermo Scientific)
Gloves
Protect extra-sensitive hands from irritation and occlusion of the skin associated with frequent gloving with Thermo Scientific™ UltraFIT™ Nylon glove liners.
  • These liners are are pre-shrunk for multiple laundering
  • Form-fitting design for a skin-tight fit
  • Ultra-thin fabric for optimum dexterity and sense of touch
  • Extra layer of insulation when working in cooler areas
  • Seamless fingertips
  • Cuffs are color coded for easy sorting
  • Nonlinting and washable
These glove liners offer added protection to individuals whose hands are extra-sensitive to the chemicals and powder contained in latex, nitrile, and vinyl gloves. Knitted liners are full-fingered and made with an ultra-thin nylon fabric, resulting in a skin-tight fit and seamless fingertips. Liners are specifically designed to fit under gloves and provide a physical barrier that helps prevent skin irritation without affecting the sense of touch. The physical barrier also protects against the occlusion of the skin associated with frequent glove use. Washable and designed for multiple use. Color-coded polyester cuff bands identify sizes. Ambidextrous. Liner color: white.