Junkin™ Safety Stretcher Bridle Sling
Supplier: Junkin
JSA-300-X
56612-685EA 478.07 USD
56612-685
Junkin™ Safety Stretcher Bridle Sling
First Aid Supplies Evacuation Equipment
For use in rescue situations where stretcher and victim need to be lifted out of danger.
  • Provides horizontal or vertical lifting capacity of up to 907 kg (2000 lbs.)
  • Made of 2.5 cm (1") nylon webbing with forged nap hooks and Int. Ø. 7.6 cm (3") forged steel suspension ring.