Fendall® Saline Eyewash Bottles, Honeywell Safety
Supplier: Honeywell Safety Products
32-000455-0000-H5 32-000451-0000 32-000452-0000
56611-124CS 322.05 USD
56611-124 56611-116 56611-118
Fendall® Saline Eyewash Bottles, Honeywell Safety
First Aid Supplies Eyewash Stations and Eyewash Bottles
This bottle contains a sterile, buffered saline solution for emergency eye care.
  • Expiry dates and lot numbers for end-user protection
  • User-controlled volume delivery: one, four, and eight-ounce products
It delivers drops or an irrigating stream of solution, depending on the squeeze pressure applied to the bottle.
Each bottle features extended flow openings and twist-off tops for easy access in an emergency.