Fendall® Porta Stream® I and Porta Stream II Eyewash Stations, Honeywell Safety
Supplier: Honeywell Safety Products
32-000200-0000
56611-102EA 324.77 USD
56611-102 56611-104 89030-548
Fendall® Porta Stream® I and Porta Stream II Eyewash Stations, Honeywell Safety
First Aid Supplies Eyewash Stations and Eyewash Bottles
The lightweight Porta Stream® eyewash stations require no plumbing and can be easily installed near workplace hazards. Stations provide uninterrupted, gravity-fed, hands-free flushing that is initiated by the removal of a nozzle strap.
  • Full flow nozzles cover a wide area around the eyes
  • Stations can be wall-mounted or placed on mobile carts or stands
  • Bright green color ensures easy identification in emergency situations
  • Sturdy, one-piece molded construction of each station features a large fill hole and illustrated operating instructions
The Porta Stream® I is a secondary eyewash device that delivers six minutes of uninterrupted flushing.
The Porta Stream® II is a primary eyewash device with a 15 minute flushing period, which fulfills ANSI Z358.1 requirements. It features a 60.5 L (16 gal.) fluid capacity and maintains a constant 1.5 L/min. (0.4 gal./min.) flow rate. Porta Stream® II eyewashes are available with saline concentrate or Fendall water preservative.
This station is supplied with 2 L (70 oz.) of saline concentrate, which provides 22.7 L (6 gal.) of solution when mixed with water.