Fendall® Flash Flood® Emergency Eyewash Stations, Honeywell Safety
Supplier: Honeywell Safety Products
32-000400-0000
56611-100EA 457.53 USD
56611-100
Fendall® Flash Flood® Emergency Eyewash Stations, Honeywell Safety
First Aid Supplies Eyewash Stations and Eyewash Bottles
The Fendall Flash Flood delivery system offers immediate, hands-free eye flushing, and is portable to meet the needs of changing work-place hazards.
  • One-gallon sealed cartridges are easy to store
  • Easy installation with no plumbing required
  • Dual nozzles provide simultaneous flushing to both eyes for three minutes
  • Highly portable system may be easily wall mounted
Unique cartridge design delivers a buffered, saline solution superior to tap water for emergency eye care. Nozzle strap is easily removed for hands-free flushing, leaving hands free to hold eyelids open. Eyesaline solution cartridges, once installed, require a 6-month maintenance cycle. Unopened cartridges have a shelf life of up to 24 months.
Can be easily wall mounted for placement near most workplace hazards.