Drum Faucet, Eagle Manufacturing
Supplier: Eagle Manufacturing
1974B
56610-550EA 307.49 USD
56610-550
Drum Faucet, Eagle Manufacturing
Faucets and Sinks Faucets
Self-closing brass faucet fits 1.9 cm (3/4") opening in drum
With internal flame arrester for safety.
Certifications: FM approved.