Wood Laminate Storage Cabinet, Justrite®
Supplier: Justrite
24140
56610-486EA 3391.6 USD
56610-486
Wood Laminate Storage Cabinet, Justrite®
Furniture Storage Cabinets
These wood laminate cabinets are suitable for highly corrosive liquids such as phenol, nitric acid and sulfuric acid.
  • Color: blue
Long lasting wood construction provides exceptional strength along with stainless handles and hinges to resist corrosion from harsh acids.