IBC Spill Containment Unit, Model 1680, Eagle Manufacturing
Supplier: Eagle Manufacturing
1680
56610-204EA 3630.9 USD
56610-204
IBC Spill Containment Unit, Model 1680, Eagle Manufacturing
Drum Storage
Made of 100% polyethylene, containment tub features a 1514L (400gal.) secondary spill capacity
The extra-wide, heavy-duty metal platform shelf accepts intermediate bulk container (IBC) units from 96.5–127cm (38–50") in width.
Certifications: Meets EPA requirement 40 CFR 264.175.