Modular Spill Containment Platforms with Grating, Eagle Manufacturing
Supplier: Eagle Manufacturing
1633 1632 1634
56609-993EA 463.16 USD
56609-993 56618-544 56618-546
Modular Spill Containment Platforms with Grating, Eagle Manufacturing
Drum Storage
Economical, low-profile platform units support 208.2L (55gal.) drums, provide incidental spill containment, and connect to form multiple configurations.
Constructed of chemical-resistant high-density polyethylene. Translucent tub is safety yellow with black grating supports. Grating has 4.4cm (13/4") square openings and removes easily for cleaning. Polyethylene ramp (56610-212) fits all platform units and simplifies drum placement.
Ordering information: One-drum platform includes one U-channel connector for joining units; Two- and four-drum platforms include two connectors.