Laboratory Safety Cans, Eagle Manufacturing
Supplier: Eagle Manufacturing
1301
56609-970EA 230 USD
56609-970
Laboratory Safety Cans, Eagle Manufacturing
Safety Cans
For safe transport and dispensing of flammable liquids.
  • Flame arrester reduces the possibility of ignition of flammable liquid vapors
  • Spout cap with nitrile rubber gasket ensures a leakproof seal
  • Electrically welded construction
  • Oval shape conserves space
  • Rolled edge pour spout
Certifications: Meet OSHA requirements and NFPA Code 30.