Type II Safety Cans, Galvanized Steel, Eagle Manufacturing
Supplier: Eagle Manufacturing
U226S U251S
56609-765EA 397.33 USD
56609-765 56609-801
Type II Safety Cans, Galvanized Steel, Eagle Manufacturing
Safety Cans
For safe transport, storage, and dispensing of flammable liquids.
Equipped with precision air vent that eliminates back pressure and permits rapid pouring. Vent opens automatically when carry/pour handle is engaged in rear groove of filler spout and pressure is exerted. Helps reduce danger of explosion when dispensing flammable liquids through the flexible metal spout into small openings. Nonsparking brass flame arresters in both the filler opening and the pour spout smother any flame at mouth of pour spout.
Flexible spout O.D.: 2.2cm (7/8"). Filler opening I.D.: 4.1cm (15/8"). Made of heavy galvanized steel.
Certifications: Meets OSHA requirements. UL listed. FM approved.