Type I Safety Cans, Justrite®
Supplier: Justrite
7110100
56609-600EA 113.23 USD
56609-600
Type I Safety Cans, Justrite®
Safety Cans
Designed for carrying flammable liquids to pour into containers with wider openings.
  • Reduce fire risks and improve safety during everyday use of flammables
  • Single spout welded to the can allows both filling and pouring
  • All cans have a flame arrester and are tested under pressure to guarantee leakproof construction
  • Spring-mounted relief vent mechanism automatically vents pressure when it reaches 5psig
This fixed handle model is equipped with a trigger release cap. Heavy-duty coated steel plate construction with red powder-coat finish protects against abrasion and rough use. Heavy steel spout is welded to dome of can to provide extra rigidity and high resistance to leak-causing nicks or dents. Spring-mounted relief vent mechanism automatically vents pressure when it reaches 5psig. Containers are tested under pressure to guarantee leakproof construction. All cans have flame arrester.
Certifications: FM approved. UL and C-UL listed.