Nonmetallic Laboratory Safety Cans, Justrite®
Supplier: Justrite
14169
56609-586EA 660.42 USD
56609-586
Nonmetallic Laboratory Safety Cans, Justrite®
Safety Cans
High-density polyethylene safety cans for use on shelves or in tilt stands have self-closing faucet for convenient, controlled dispensing of hazardous liquids. Fill spout has self-closing pressure relief cap and double mesh flame arrester. The 18.9L (5gal.) cans have built-in vacuum breakers. Oval-shaped safety can 56609-586 has stainless steel faucet and round can 56609-589 has brass faucet, both mounted 5cm (2") above bottom. Tilt can 56609-588 with top faucet rests on a sturdy metal cradle which measures 36.8x35.5cm (14½x14") at the base.
  • Feature additional self-closing faucet that allows safe dispensing of hazardous liquids into test tubes, small flasks, and beakers
  • Fill spout has self-closing pressure relief cap and double mesh flame arrester
  • Current-carrying carbon insert imbedded in the can provides proper grounding
  • 5cm (2") with faucet mounted above the bottom allows dispensing without lifting
  • 18.9L (5gal.) cans have built-in vacuum breaker
  • FM approved