Respirator Cartridges and Filters, Honeywell Safety
Supplier: Honeywell Safety Products
7580P100 7506N95 75SCL 7531N95 N750035 N75002L N75003L N75004L 75SCL 7581P100L 7582P100L 7583P100L 7584P100L 75SCP100L
56222-964CS 1170.69 USD
56222-964 56222-965 56223-250 56222-972 56223-268 89400-380 89400-382 89400-384 89400-386 89400-390 89400-392 89400-394 89400-396 89400-398
Respirator Cartridges and Filters, Honeywell Safety
For use with North Safety Products air purifying dual cartridge respirators (see 56222-944 series).
  • Simplify your inventory with one line of cartridges and filters
  • NIOSH approved when used with Nort Safety Products 7700, 7600, 5500, and 5400 air-purifying respirators
Choose from a wide variety of cartridges, filters, and cartridge/filter combinations to suit your applications. Adapter assembly 56223-268 attaches pancake filter 56223-266 to a gas and vapor cartridge, and attaches to chemical cartridges except for the Defender® multipurpose cartridge.
All cartridges and filters are NIOSH approved (meet 42 CFR 84 requirements) when used with North Safety products air-purifying respirators.