8214 N95 Respirator, 3M™
Supplier: 3M
7000002083
56222-628CS 1444.63 USD
56222-628
8214 N95 Respirator, 3M™
Respirators Disposable Respirators Particulate Respirators
This particulate respirator is designed for applications such as welding, soldering and other operations where metal fumes may be present including those with ozone and faceseal and nuisance level organic vapors.
  • Used in applications involving metal fumes
  • Individual packaging designed for vending
  • Advanced electrostatic media
  • Adjustable buckle straps
  • Cake resistant filter media
Carbon filter media reduces exposure to ozone/nuisance level of organic vapors.