Attest™ Biological Indicators for EO, 3M™
Supplier: Healthcare Products
MMM 1264
56222-092BX 176.95 USD
56222-092 56222-516
Attest™ Biological Indicators for EO, 3M™
Sterilization Indicators
3M™ Attest™ self-contained BI is designed for monitoring ethylene oxide (EO) sterilization processes.
It consists of a Bacillus atrophaeus spore strip; sealed glass ampule with growth medium and bromothymol blue pH indicator dye system; green color-coded cap with holes for sterilant penetration and a filter as a bacterial barrier, and a chemical indicator on the label that changes to green when processed. After sterilization, the vial is 'crushed' to join the growth media with the processed spore strip. The BI is incubated for 48 hours for a visual color change readout. A color change to yellow indicates surviving spores and a positive result.