2310 Disposable N99 Premium Particulate Respirator, Moldex®
Supplier: Moldex/Metric
2310
56219-030CS 840.86 USD
56219-030
2310 Disposable N99 Premium Particulate Respirator, Moldex®
Respirators Disposable Respirators Particulate Respirators
Provides a filter efficiency level of 99% or greater against particulate aerosols free of oil up to 10 times Permissible Exposure Limit (PEL).
  • Exhale valve lets workers breathe easily and stay cool
  • Contoured facepiece features a soft-spun linerSoft foam nose seal fits easily and comfortably without a metal noseband
  • The polymesh shell resists collapsing, even in heat and humidity and keeps the filter clean-looking longer
  • Straps are securely attached so they do not break at the seal
Suitable for most welding fumes. Fits easily under welding hoods. Flame retardants added to the Dura-Mesh® shell.
Caution: User is responsible for checking current OSHA regulations and applicable guidelines.