2500 Disposable N95 Particulate Respirators, Moldex®
Supplier: Moldex/Metric
2500N95
56219-028CS 1376.75 USD
56219-028
2500 Disposable N95 Particulate Respirators, Moldex®
Respirators Disposable Respirators Particulate Respirators
Used when oil aerosols are not present in industries such as aluminum reduction, paper mills, coal-fired utility plants, petrochemicals, chemical processing and glass etching
  • Naturally contoured for a comfortable, secure fit
  • Seals easily and without a metal noseband
  • Durable, crush-resistant polymesh outer shell keeps the filter cleaner-looking longer
  • Resists collapsing, even in heat and humidity
  • Exhale valve reduces breathing resistance and hot air build up
  • Disposable
Provides a filter efficiency level of 95% or greater against particulate aerosols free of oil up to 10 times Permissible Exposure Limit (PEL). For relief from nuisance levels (less than OSHA PEL) of acid gas irritants.