2400 Disposable N95 Particulate Respirators, Moldex®
Supplier: Moldex/Metric
2400N95
56219-026CS 1470.77 USD
56219-026
2400 Disposable N95 Particulate Respirators, Moldex®
Respirators Disposable Respirators Particulate Respirators
Designed for use against nuisance levels (less than OSHA Permissible Exposure Limit) of organic vapor odors and/or ozone
  • Provides a filter efficiency level of 95% or greater against particulate aerosols free of oil
  • Suitable for most welding fumes, and fits easily under welding hoods
  • Flame-retardants added to the Dura-Mesh® shell
  • Workers stay cool with an exhale valve that reduces breathing resistance and hot air build-up
  • Facepiece is naturally contoured without pressure points
  • Dura-Mesh® shell resists collapsing, even in heat and humidity
Workers stay cool with an exhale valve that reduces breathing resistance and hot air build-up. Facepiece is naturally contoured without pressure points. Dura-Mesh® shell resists collapsing, even in heat and humidity. Stays cleaner-looking longer.
Suitable for most welding fumes. Fits easily under welding hoods. Flame-retardants added to the Dura-Mesh® shell. (Note: user is responsible for checking current OSHA regulations and applicable guidelines for welding applications.)