2200 Disposable N95 Particulate Respirators, Moldex®
Supplier: Moldex/Metric
2200N95
56219-004CS 837.17 USD
56219-004 56219-010
2200 Disposable N95 Particulate Respirators, Moldex®
Respirators Disposable Respirators Particulate Respirators
Disposable respirators provide a filter efficiency level of 95% or greater against particulate aerosols free of oil up to 10 times OSHA Permissible Exposure Limit (PEL).
  • Facepiece is naturally contoured without pressure points
  • Seal is maintained without a metal noseband
  • Straps are securely attached so they do not break at the seal
  • Dura-Mesh® outer shell resists collapsing in heat and humidity and stays clean-looking longer
Available in three sizes: small, medium/large, and alternate shape (for workers with rounder facial features and flatter nose bridges).