Nunc® CryoTube® Vial Rack, Thermo Scientific
Supplier: Thermo Fisher Scientific
376589
55710-262CS 836.84 USD
55710-262
Nunc® CryoTube® Vial Rack, Thermo Scientific
Racks Cryogenic Vial Racks
Rack holds externally or internally threaded CryoTube® vials
The rack, manufactured from polyphenylene oxide, allows one-handed operation when used with starfoot tubes.