Barnstead/Thermolyne Locator® Cryobiological Storage System Boxes, Thermo Scientific
Supplier: Thermo Fisher Scientific
CS509X3
55709-903CS 1112.5 USD
55709-903
Barnstead/Thermolyne Locator® Cryobiological Storage System Boxes, Thermo Scientific
Boxes Cryoboxes
Nalgene® Cryoplastic boxes are of durable, ultra-low temperature polycarbonate construction
One-way lid reduces the chances of sample spillage. A printed grid system on the lid facilitates easy logging and retrieval of samples.