Chart Paper, Thermo Scientific
Supplier: VWR International
90520H11
55702-630PK 120.64 USD
55702-630
Chart Paper, Thermo Scientific
Paper
Circular chart paper, non-thermal.
  • Pressure-sensitive
  • 7 days
Range: −40...+25 °C