Laboratory Pumps, Oil-Less Reciprocating, Gast®
Supplier: Gast Manufacturing
1HAB-25-M100X
54907-086EA 4244.75 USD
54907-086
Laboratory Pumps, Oil-Less Reciprocating, Gast®
Pumps Vacuum Pumps
Special purpose unit functions as both vacuum pump and compressor
Features include oil-less operation, low maintenance and rugged construction. The motor is a single phase, 1/6 hp that delivers 1725rpm.
Ordering information: Complete with gauges, hose barbs, regulators, cord with on/off switch, and carrying handle.